Boc-Trp-OMe 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Boc-L-tryptophan methyl ester
A248673-500G
A248673-5G
A248673-10G
A248673-100G
103851-670EA
9
USD
103851-670
76692-420
77304-458
77568-410
77877-836
Boc-Trp-OMe 98%
Boc-Trp-OMe
Precaución: For research use only.
Formula:
C₁₇H₂₂N₂O₄ Boiling Pt: 505.7±45.0 °C (760 mm Hg) Melting Pt: 147…149 °C Flash Pt: 259.7 °C Storage Temperature: Freezer |
MDL Number:
MFCD01075094 CAS Number: 33900-28-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...