N(α)-Boc-L-tryptophan 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Trp-OH , Boc-Trp-OH

A191954-25G A191954-100G A191954-10G A191954-500G
103847-014EA 7 USD
103847-014 77084-882 77274-452 77274-454 77748-226
N(α)-Boc-L-tryptophan 97%
N(α)-Boc-L-tryptophan

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₄
MW: 304.3 g/mol
Boiling Pt: 535.7 °C (760 mm Hg)
Melting Pt: 136 °C
Flash Pt: 277.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00065595
CAS Number: 13139-14-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR