N-ɑ-Boc-N(in)-formyl-L-tryptophan 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Boc-1-formyl-L-tryptophan, N-ɑ-(tert-Butoxycarbonyl)-N1-formyl-L-tryptophan, N-ɑ-Boc-N1-formyl-L-tryptophan, Boc-Trp(For)-OH, Boc-Trp(CHO)-OH
A170488-100G
A170488-10G
A170488-1G
A170488-5G
103806-506EA
10
USD
103806-506
103844-780
76973-982
76973-984
77081-890
77081-892
N-ɑ-Boc-N(in)-formyl-L-tryptophan 95%
N-alpha-Boc-N(in)-formyl-L-tryptophan
Precaución: for Research Use Only
Formula:
C₁₇H₂₀N₂O₅ MW: 332.36 g/mol Melting Pt: 100 °C Storage Temperature: Freezer |
MDL Number:
MFCD00065992 CAS Number: 47355-10-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...