N-(tert-Butoxycarbonyl)-L-tert-leucine 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-tert-leucine, Boc-Tle-OH, (S)-2-(Boc-amino)-3,3-dimethylbutyric Acid, (S)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid

A124684-1000G A124684-100G A124684-10G A124684-25G A124684-500G A124684-5G
76810-764EA 294.94 USD
76810-764 76810-766 76810-768 76810-770 76810-772 77298-016
N-(tert-Butoxycarbonyl)-L-tert-leucine 98%
N-(tert-Butoxycarbonyl)-L-tert-leucine

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
Boiling Pt: 350 °C (760 mm Hg)
Melting Pt: 118…121 °C
Flash Pt: 165.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00065574
CAS Number: 62965-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR