Boc-Tic-OH 98%

Supplier: Ambeed
Warning

Synonyms: (S)-N-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-Tic-OH, (S)-2-(tert-Butoxycarbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, Boc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (S)-2-Boc-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

A238609-25G A238609-100G A238609-10G A238609-500G A238609-1G A238609-5G
103811-238EA 5 USD
103811-238 103851-024 77090-148 77275-960 77275-962 77275-964 77304-114 77304-116
Boc-Tic-OH 98%
Boc-Tic-OH

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₄
MW: 277.32 g/mol
Boiling Pt: 266.6 °C (760 mm Hg)
Melting Pt: 59…63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00143845
CAS Number: 78879-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR