N-Boc-L-threonine methyl ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Thr-OMe, (2S,3R)-Methyl 2-((tert-butoxycarbonyl)amino)-3-hydroxybutanoate
A265648-25G
A265648-5G
A265648-100G
A265648-500G
A265648-1G
103812-742EA
9
USD
103812-742
103852-638
77092-338
77092-340
77113-788
77113-790
77113-792
77304-922
N-Boc-L-threonine methyl ester 95%
N-Boc-L-threonine methyl ester
Precaución: for Research Use Only
Formula:
C₁₀H₁₉NO₅ MW: 233.264 g/mol Boiling Pt: 206 °C (760 mm Hg) Flash Pt: 172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00270479 CAS Number: 79479-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...