Boc-Thr(tBu)-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,3R)-3-(tert-Butoxy)-2-((tert-butoxycarbonyl)amino)butanoic acid, N-Boc-O-tert-butyl-L-threonine
A195018-100G
A195018-10G
A195018-1G
A195018-5G
103816-714EA
9
USD
103816-714
103847-328
76721-260
76721-262
77085-276
77085-278
Boc-Thr(tBu)-OH 97%
Boc-Thr(tBu)-OH
Precaución: for Research Use Only
Formula:
C₁₃H₂₅NO₅ Boiling Pt: 391.3 °C (760 mm Hg) Melting Pt: 95…98 °C Flash Pt: 190.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151115 CAS Number: 13734-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...