N-Boc-O-methyl-L-threonine 96%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-2-(Boc-amino)-3-methoxybutyric acid, (2S,3R)-2-((tert-Butoxycarbonyl)amino)-3-methoxybutanoic acid, Boc-Thr(Me)-OH

A207803-10G A207803-1G A207803-100G
103817-828EA 29 USD
103817-828 103848-514 76691-354 77086-800 77600-682
N-Boc-O-methyl-L-threonine 96%
N-Boc-O-methyl-L-threonine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₅
MW: 233.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00076990
CAS Number: 48068-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR