Boc-Ser-Otbu 95%

Supplier: Ambeed
Warning

Synonyms: Boc-Ser-OtBu

A298567-250MG A298567-1G
103814-186EA 49 USD
103814-186 103835-332 77114-418 77305-742
Boc-Ser-Otbu 95%
Boc-Ser-Otbu

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₅
MW: 261.318 g/mol
Boiling Pt: 381.9±32.0 °C (760 mm Hg)
Flash Pt: 184.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00190832
CAS Number: 7738-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR