N-(tert-Butoxycarbonyl)-L-serinemethylester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-Ser-OMe, N-Boc-L-serine Methyl Ester, BOC-L-Serine methyl ester
A164821-100G
A164821-500G
A164821-1000G
A164821-25G
A164821-5G
103808-948EA
315
USD
103808-948
103844-166
76997-984
76997-986
77320-354
77320-356
77320-358
N-(tert-Butoxycarbonyl)-L-serinemethylester 97%
N-(tert-Butoxycarbonyl)-L-serine methyl ester
Precaución: for Research Use Only
Formula:
C₉H₁₇NO₅ MW: 219.24 g/mol Boiling Pt: 354.3 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00191869 CAS Number: 2766-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...