(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-serine benzyl ester, Boc-Ser-OBzl, BOC-L-Serine benzyl ester

A244097-1G A244097-25G A244097-5G A244097-100G A244097-10G A244097-500G
103811-568EA 6 USD
103811-568 103851-398 77090-606 77090-608 77090-610 77113-290 77113-292 77113-294
(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 98%
(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₅
MW: 295.3 g/mol
Boiling Pt: 458.8 °C (760 mm Hg)
Melting Pt: 91 °C
Flash Pt: 231.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00076983
CAS Number: 59524-02-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR