(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Homopro-OH, 1-(tert-Butoxycarbonyl)-L-pipecolic Acid, Boc-HomoPro-OH, 1-Boc-L-pipecolic Acid, (S)-1-Boc-2-piperidinecarboxylic Acid, Boc-Pip-OH, (S)-N-BOC-Piperidine-2-carboxylic acid

A312966-100G A312966-10G A312966-500G A312966-25G A312966-5G
103835-900EA 6 USD
103835-900 76802-770 76802-772 76802-774 76838-718 76838-720
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid 98%
(S)-1-(tert-Butoxycarbonyl)piperidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₉NO₄
Melting Pt: 122…126 °C
Flash Pt: 167.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151904
CAS Number: 26250-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR