N-(tert-Butoxycarbonyl)-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-phenylalanine, BOC-L-Phenylalanine, N-α-t-Boc-L-phenylalanine, Boc-Phe-OH

A161128-100G A161128-10G A161128-500G A161128-25G
103808-892EA 147 USD
103808-892 77273-302 77273-304 77273-306 77300-516
N-(tert-Butoxycarbonyl)-L-phenylalanine 98%
N-(tert-Butoxycarbonyl)-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
Boiling Pt: 426.6 °C (760 mm Hg)
Melting Pt: 85…87 °C
Flash Pt: 211.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00002663
CAS Number: 13734-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR