(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-N-Boc-L-phenylalanine, Boc-p-amino-Phe-OH, Boc-Phe(4-NH2)-OH

A161133-100G A161133-10G A161133-250MG A161133-1G A161133-25G A161133-5G
103805-456EA 13 USD
103805-456 103843-694 76783-854 76783-856 76783-858 77319-658 77319-660 77319-662
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 98%
(S)-3-(4-Aminophenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₀N₂O₄
MW: 280.3 g/mol
Boiling Pt: 484.9 °C (760 mm Hg)
Flash Pt: 247 °C
Storage Temperature: Ambient
MDL Number: MFCD00038139
CAS Number: 55533-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR