N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Phe(4-F)-OH, N-Boc-4-fluoro-L-phenylalanine

A128946-100G A128946-250MG A128946-25G A128946-1G A128946-5G A128946-10G
103801-206EA 15 USD
103801-206 77190-000 77190-002 77298-326 77313-560 77313-562 77745-930
N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine 98%
N-(tert-Butoxycarbonyl)-4-fluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₄
MW: 283.30 g/mol
Boiling Pt: 431.2 °C (760 mm Hg)
Melting Pt: 80 °C
Flash Pt: 214.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079672
CAS Number: 41153-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR