Boc-Phe(4-CN)-OH 95%

Supplier: Ambeed
Warning

Synonyms: Boc-4-cyano-Phe-OH, N-Boc-4-cyano-L-phenylalanine, Boc-Phe(4-CN)-OH, N-(tert-Butoxycarbonyl)-4-cyano-L-phenylalanine

A364670-100G A364670-250MG A364670-1G A364670-10G
103826-364EA 22 USD
103826-364 103837-384 76788-596 76788-598 76840-614 77668-690
Boc-Phe(4-CN)-OH 95%
Boc-Phe(4-CN)-OH

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
Boiling Pt: 494 °C (760 mm Hg)
Melting Pt: 152.3 °C
Flash Pt: 252.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00672526
CAS Number: 131724-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR