N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: N-Boc-3-fluoro-L-phenylalanine, Boc-Phe(3-F)-OH

A229397-10G A229397-250MG A229397-1G A229397-25G A229397-5G A229397-100G
103819-474EA 6 USD
103819-474 103850-374 76786-200 76786-202 77089-216 77089-218 77089-220 77872-165
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine 97%
N-(tert-Butoxycarbonyl)-3-fluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₄
Boiling Pt: 425 °C (760 mm Hg)
Melting Pt: 75…80 °C
Flash Pt: 210.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00672522
CAS Number: 114873-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR