N-Boc-3-cyano-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Phe(3-CN)-OH

A145209-250MG A145209-25G A145209-5G
103803-504EA 12 USD
103803-504 103841-586 76783-122 76783-124 77299-436
N-Boc-3-cyano-L-phenylalanine 97%
N-Boc-3-cyano-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₅H₁₈N₂O₄
MW: 290.319 g/mol
Boiling Pt: 308.407 °C (760 mm Hg)
Melting Pt: 124.1 °C
Flash Pt: 140.32 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00797560
CAS Number: 131980-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR