Boc-3-chloro-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-L-phenylalanine, N-BOC protected, Boc-Phe(3-Cl)-OH

A136708-10G A136708-250MG A136708-5G A136708-1G A136708-25G
103802-336EA 25 USD
103802-336 76782-700 76782-702 77298-862 77315-258 77315-260
Boc-3-chloro-L-phenylalanine 97%
Boc-3-chloro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈ClNO₄
MW: 299.76 g/mol
Boiling Pt: 452.5 °C (760 mm Hg)
Melting Pt: 110 °C
Flash Pt: 227.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672515
CAS Number: 114873-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR