Boc-3-bromo-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Phe(3-Br)-OH

A155273-10G A155273-250MG A155273-1G A155273-5G A155273-100G A155273-500G
103804-738EA 7 USD
103804-738 103842-932 76783-546 76783-548 77318-652 77318-654 77600-216 77871-770
Boc-3-bromo-L-phenylalanine 98%
Boc-3-bromo-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₄
Boiling Pt: 475.3 °C (760 mm Hg)
Melting Pt: 106.1 °C
Flash Pt: 241.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01317710
CAS Number: 82278-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR