N-Boc-2-fluoro-L-phenylalanine 97%

Supplier: Ambeed
Warning

A410775-1G A410775-25G A410775-5G A410775-10G A410775-250MG
103827-218EA 10 USD
103827-218 103838-266 76841-790 76841-792 76841-794 77196-072 77196-074
N-Boc-2-fluoro-L-phenylalanine 97%
N-Boc-2-fluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₄
Boiling Pt: 427.3 °C (760 mm Hg)
Melting Pt: 92…98 °C
Flash Pt: 212.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672520
CAS Number: 114873-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR