(S)-N-Boc-2-bromophenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Phe(2-Br)-OH

A288683-100G A288683-1G A288683-5G A288683-25G
103834-946EA 20 USD
103834-946 76787-500 76787-502 76837-512 77305-536
(S)-N-Boc-2-bromophenylalanine 97%
(S)-N-Boc-2-bromophenylalanine

Precaución: for Research Use Only

Formula: C₁₄H₁₈BrNO₄
MW: 344.205 g/mol
Boiling Pt: 471.3 °C (760 mm Hg)
Melting Pt: 139…140 °C
Flash Pt: 238.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01317709
CAS Number: 261165-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR