Boc-Orn-OH 97%

Supplier: Ambeed
Warning

Synonyms: Nα-Boc-L-ornithine

A164963-10G A164963-1G A164963-100G
103805-922EA 5 USD
103805-922 103844-180 76998-002 77320-376 77671-324
Boc-Orn-OH 97%
Boc-Orn-OH

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₄
Boiling Pt: 398.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00076970
CAS Number: 21887-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR