Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-Orn(Z)-OH, Nɑ-Boc-Nδ-Cbz-L-ornithine, Nδ-Benzyloxycarbonyl-Nα-Boc-L-ornithine

A622812-25G A622812-5G A622812-100G A622812-500G A622812-1G
103840-226EA 6 USD
103840-226 76844-392 76844-394 77035-266 77035-268 77307-982
Nɑ-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine 98%
Nalpha-(tert-Butoxycarbonyl)-Ndelta-carbobenzoxy-L-ornithine

Precaución: for Research Use Only

Formula: C₁₈H₂₆N₂O₆
MW: 366.41 g/mol
Boiling Pt: 579.1 °C (760 mm Hg)
Melting Pt: 94 °C at N/A mmHg
Flash Pt: 304 °C
Storage Temperature: Ambient
MDL Number: MFCD00038259
CAS Number: 2480-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR