Boc-N-Me-Val-OH 97%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butoxycarbonyl)-N-methyl-L-valine, N-Boc-N-methyl-L-valine, Boc-N-Me-Val-OH

A148990-100G A148990-10G A148990-250MG A148990-500G A148990-25G A148990-5G A148990-1G
103803-992EA 6 USD
103803-992 103842-112 76798-546 76798-548 76798-550 76798-552 77299-700 77299-702 77317-588
Boc-N-Me-Val-OH 97%
Boc-N-Me-Val-OH

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.292 g/mol
Boiling Pt: 322.4 °C (760 mm Hg)
Flash Pt: 148.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00038760
CAS Number: 45170-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR