Boc-N-Me-Phe-OH 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-N-methyl-L-phenylalanine

A239099-1G A239099-25G A239099-5G A239099-10G
103811-266EA 26 USD
103811-266 103851-064 77090-200 77090-202 77090-204 77275-966
Boc-N-Me-Phe-OH 95%
Boc-N-Me-Phe-OH

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MW: 279.336 g/mol
Boiling Pt: 405 °C (760 mm Hg)
Flash Pt: 198.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038754
CAS Number: 37553-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR