Boc-N-methyl-L-leucine 95%

Supplier: Ambeed
Warning

Synonyms: Boc-N-Me-Leu-OH

A125067-100G A125067-10G A125067-25G A125067-1G
103800-618EA 6 USD
103800-618 76688-334 76688-336 76688-338 76688-340 77312-768
Boc-N-methyl-L-leucine 95%
Boc-N-methyl-L-leucine

Precaución: for Research Use Only

Formula: C₁₂H₂₃NO₄
MW: 245.319 g/mol
Boiling Pt: 338.2 °C (760 mm Hg)
Melting Pt: 55…60 °C
Flash Pt: 158.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038522
CAS Number: 53363-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR