N-(tert-Butoxycarbonyl)-N-methyl-D-alanine 97%

Supplier: Ambeed
Warning

Synonyms: BOC-N-methyl-D-alanine, (R)-2-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid, Boc-N-Me-D-Ala-OH, N-Boc-N-methyl-D-alanine

A211793-100G A211793-10G A211793-250MG A211793-1G A211793-25G A211793-5G
103818-138EA 8 USD
103818-138 103848-834 76691-514 76691-516 76691-518 77087-258 77087-260 77087-262
N-(tert-Butoxycarbonyl)-N-methyl-D-alanine 97%
N-(tert-Butoxycarbonyl)-N-methyl-D-alanine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.24 g/mol
Boiling Pt: 296.3 °C (760 mm Hg)
Melting Pt: 90…94 °C
Flash Pt: 133 °C
Storage Temperature: Ambient
MDL Number: MFCD00063137
CAS Number: 19914-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR