Boc-N-methyl-D-alanine 98%

Supplier: Ambeed
Warning

Synonyms: Boc-N-Me-Ala-OH, N-Boc-N-methyl-L-alanine

A139244-100G A139244-10G A139244-1G A139244-25G A139244-5G
103802-716EA 5 USD
103802-716 77272-410 77272-412 77299-060 77299-062 77315-798
Boc-N-methyl-D-alanine 98%
Boc-N-methyl-D-alanine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MW: 203.2 g/mol
Boiling Pt: 296.3 °C (760 mm Hg)
Melting Pt: 88…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00037242
CAS Number: 16948-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR