Boc-N-2-dimethylalanine 97%

Supplier: Ambeed
Warning

Synonyms: N-BOC-N,2-Dimethylalanine, Boc-N-Me-Aib-OH

A251967-10G A251967-250MG A251967-1G
103812-016EA 23 USD
103812-016 103851-858 76692-488 76692-490 77091-258
Boc-N-2-dimethylalanine 97%
Boc-N-2-dimethylalanine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.265 g/mol
Boiling Pt: 306.4 °C (760 mm Hg)
Melting Pt: 154…158 °C
Flash Pt: 139.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862912
CAS Number: 146000-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR