Boc-MeTyr-OH 97%

Supplier: Ambeed
Warning

A183655-100MG A183655-250MG A183655-1G A183655-5G
103815-654EA 164 USD
103815-654 103846-218 76872-842 76872-844 77301-842 77301-844
Boc-MeTyr-OH 97%
Boc-MeTyr-OH

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₅
MW: 295.33 g/mol
Boiling Pt: 626.2 °C (760 mm Hg)
Flash Pt: 332.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06658544
CAS Number: 82038-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR