Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Nepsilon-Benzyloxycarbonyl-Nα-Boc-L-lysine, Boc-Lys(Z)-OH, Nɑ-Boc-Nepsilon-Cbz-L-lysine, Boc-Lys(Cbz)-OH
A156665-100G
A156665-500G
A156665-5G
A156665-25G
A156665-1000G
103804-910EA
6
USD
103804-910
103843-112
77273-148
77273-150
77273-152
77300-206
77318-908
77748-067
Nɑ-(tert-Butoxycarbonyl)-N-ε-carbobenzoxy-L-lysine 98%
Nalpha-(tert-Butoxycarbonyl)-N-epsilon-carbobenzoxy-L-lysine
Precaución: for Research Use Only
Formula:
C₁₉H₂₈N₂O₆ MW: 380.44 g/mol Boiling Pt: 587 °C (760 mm Hg) Flash Pt: 308.8 °C Storage Temperature: Freezer |
MDL Number:
MFCD00065584 CAS Number: 2389-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...