Boc-Lys(Fmoc)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-epsilon-FMOC-N-alpha-BOC-L-Lysine

A178863-100G A178863-10G A178863-500G A178863-1G A178863-25G A178863-5G
103807-378EA 6 USD
103807-378 103845-718 77287-094 77287-096 77287-098 77301-578 77301-580 77301-582
Boc-Lys(Fmoc)-OH 98%
Boc-Lys(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Boiling Pt: 685.7 °C (760 mm Hg)
Melting Pt: 93…128 °C
Flash Pt: 368.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00062032
CAS Number: 84624-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR