N-(tert-Butoxycarbonyl)-L-serine-β-lactone 97%

Supplier: Ambeed
Warning

Synonyms: (S)-ɑ-tert-Butoxycarbonylamino-β-propiolactone, tert-Butyl (S)-(2-oxooxetan-3-yl)carbamate, N-Boc-L-serineβ-Lactone, (S)-ɑ-Boc-amino-β-propiolactone, (S)-3-(tert-Butoxycarbonylamino)-2-oxetanone

A358497-250MG A358497-25G
103826-210EA 28 USD
103826-210 103837-216 76724-552 77306-690
N-(tert-Butoxycarbonyl)-L-serine-β-lactone 97%
N-(tert-Butoxycarbonyl)-L-serine-beta-lactone

Precaución: for Research Use Only

Formula: C₈H₁₃NO₄
MW: 187.20 g/mol
Boiling Pt: 319 °C (760 mm Hg)
Melting Pt: 120…124 °C
Storage Temperature: Freezer
MDL Number: MFCD01318414
CAS Number: 98541-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR