Boc-L-pyroglutamic acid methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylate, Methyl N-Boc-L-pyroglutamate, Methyl N-(tert-Butoxycarbonyl)-L-pyroglutamate, Boc-Pyr-OMe, (S)-1-(tert-Butoxycarbonyl)-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-Boc-L-pyroglutamic acid methyll ester, (S)-1-Boc-5-oxopyrrolidine-2-carboxylic acid methyll ester, N-(tert-Butoxycarbonyl)-L-pyroglutamic acid methyll ester, 1-(tert-Butyl) 2-methyl (S)-5-oxopyrrolidine-1,2-dicarboxylate, Methyl (S)-1-Boc-5-oxopyrrolidine-2-carboxylate

A534338-25G A534338-5G A534338-100G A534338-10G A534338-1G A534338-500G
103839-706EA 6 USD
103839-706 76843-722 76843-724 77004-974 77004-976 77004-978 77004-980
Boc-L-pyroglutamic acid methyl ester 97%
Boc-L-pyroglutamic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇NO₅
MW: 243.26 g/mol
Melting Pt: 69…74 °C
Flash Pt: 172.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06809720
CAS Number: 108963-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR