N-Boc-L-leucine methyl ester 97%
Supplier: Ambeed
|
Warning
|
A195456-5G
A195456-100G
A195456-10G
A195456-500G
A195456-25G
103816-752EA
5
USD
103816-752
103847-366
77085-312
77111-944
77111-946
77111-948
77302-464
77302-466
N-Boc-L-leucine methyl ester 97%
N-Boc-L-leucine methyl ester
Precaución: for Research Use Only
Formula:
C₁₂H₂₃NO₄ Boiling Pt: 322.5 °C (760 mm Hg) Flash Pt: 113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00076952 CAS Number: 63096-02-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...