N-Boc-L-isoleucine-N-succinimidyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Boc-Ile-OSu

A109146-100G A109146-10G
103797-452EA 5 USD
103797-452 103823-244 76765-668 76765-670
N-Boc-L-isoleucine-N-succinimidyl ester 97%
N-Boc-L-isoleucine-N-succinimidyl ester

Precaución: for Research Use Only

Formula: C₁₅H₂₄N₂O₆
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00022585
CAS Number: 3392-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR