(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid 97%

Supplier: Ambeed
Warning

A225331-100G A225331-10G A225331-25G A225331-1G A225331-5G
103819-172EA 6 USD
103819-172 103850-066 76786-112 76786-114 77088-754 77303-738 77303-740
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid 97%
(S)-2-((tert-Butoxycarbonyl)amino)-4-phenylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
Melting Pt: 76…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00076904
CAS Number: 100564-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR