N(α)-Boc-L-histidine 98%

Supplier: Ambeed
Warning

A246244-5G A246244-100G A246244-500G A246244-25G A246244-10G
103811-700EA 8 USD
103811-700 103851-526 77090-816 77276-186 77276-188 77304-378 77568-394
N(α)-Boc-L-histidine 98%
N(α)-Boc-L-histidine

Precaución: For research use only.

Formula: C₁₁H₁₇N₃O₄
Boiling Pt: 522.2 °C (760 mm Hg)
Melting Pt: 196…199 °C
Flash Pt: 269.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00065576
CAS Number: 17791-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR