N-Boc-3-benzyloxymethyl-L-histidine 97%

Supplier: Ambeed
Warning

Synonyms: Boc-His(Bom)-OH, N-[(1,1-Dimethylethoxy)carbonyl]-3-[(phenylmethoxy)methyl]- L-histidine, Boc-His(3-Bom)-OH

A104551-10G A104551-250MG A104551-25G A104551-1G A104551-5G
103796-730EA 8 USD
103796-730 103822-448 76809-500 76809-502 76809-504 77283-242 77309-312
N-Boc-3-benzyloxymethyl-L-histidine 97%
N-Boc-3-benzyloxymethyl-L-histidine

Precaución: for Research Use Only

Formula: C₁₉H₂₅N₃O₅
MW: 375.42 g/mol
Melting Pt: 122…125 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00043133
CAS Number: 79950-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR