Boc-glycine-N-hydroxysuccinimide ester 97%

Supplier: Ambeed
Warning

A114700-25G A114700-10G A114700-100G
103824-296EA 18 USD
103824-296 77566-026 77570-842 77623-956
Boc-glycine-N-hydroxysuccinimide ester 97%
Boc-glycine-N-hydroxysuccinimide ester

Precaución: For research use only. Inert atmosphere, store in freezer, under –20 °C.

Formula: C₁₁H₁₆N₂O₆
Melting Pt: 165…167 °C
Storage Temperature: Freezer
MDL Number: MFCD00042761
CAS Number: 3392-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR