Boc-glycine methyl ester 97%
Supplier: Ambeed
|
Warning
|
A163774-100G
A163774-10G
A163774-500G
A163774-25G
A163774-5G
A163774-1000G
103844-028EA
8
USD
103844-028
77096-228
77096-230
77096-232
77320-138
77320-140
77685-339
Boc-glycine methyl ester 97%
Boc-glycine methyl ester
Precaución: for Research Use Only
Formula:
C₈H₁₅NO₄ Boiling Pt: 266.8 °C (760 mm Hg) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038267 CAS Number: 31954-27-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...