2-(tert-Butoxycarbonylamino)-1-ethanol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(2-Hydroxyethyl)carbamic Acid tert-Butyl Ester, 2-(tert-Butoxycarbonyl)ethanolamine, N-(tert-Butoxycarbonyl)ethanolamine, 2-Boc-ethanolamine, tert-Butyl N-(2-Hydroxyethyl)carbamate, 2-(Boc-amino)-1-ethanol, Boc-Glycinol
A107162-100G
A107162-10G
A107162-25G
A107162-500G
A107162-5G
A107162-1000G
103807-962EA
217
USD
103807-962
103822-902
76765-540
76765-542
76765-544
76765-546
77283-554
77309-766
2-(tert-Butoxycarbonylamino)-1-ethanol 98%
2-(tert-Butoxycarbonylamino)-1-ethanol
Precaución: for Research Use Only
Formula:
C₇H₁₅NO₃ MW: 161.20 g/mol Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00056657 CAS Number: 26690-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...