Boc-Glu-OtBu 98%

Supplier: Ambeed
Warning

Synonyms: N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-tert-Butyl Ester, N-Boc-L-glutamic Acid 1-tert-Butyl Ester, 1-tert-Butyl N-Boc-L-glutamate, Boc-Glu-OtBu

A297281-25G A297281-5G A297281-100G A297281-10G A297281-1G A297281-1000G
103814-128EA 6 USD
103814-128 103835-270 76837-954 76837-956 77277-264 77277-266 77305-702 77748-576
Boc-Glu-OtBu 98%
Boc-Glu-OtBu

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₆
MW: 303.36 g/mol
Boiling Pt: 449.8 °C (760 mm Hg)
Flash Pt: 225.8 °C
Storage Temperature: Freezer
MDL Number: MFCD00038273
CAS Number: 24277-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR