1-Benzyl-N-(tert-butoxycarbonyl)-L-glutamate 97%

Supplier: Ambeed
Warning

Synonyms: 1-Benzyl N-Boc-L-glutamate, N-(tert-Butoxycarbonyl)-L-glutamic Acid 1-Benzyl Ester, N-Boc-L-glutamic Acid 1-Benzyl Ester, Boc-Glu-OBzl

A138972-100G A138972-10G A138972-1G A138972-5G A138972-25G
103802-682EA 9 USD
103802-682 77285-644 77285-646 77299-034 77299-036 77315-748
1-Benzyl-N-(tert-butoxycarbonyl)-L-glutamate 97%
1-Benzyl-N-(tert-butoxycarbonyl)-L-glutamate

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₆
Boiling Pt: 522.6 °C (760 mm Hg)
Melting Pt: 95…99 °C
Flash Pt: 269.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00065568
CAS Number: 30924-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR