Boc-Glu(OtBu)-OH 97%

Supplier: Ambeed
Warning

A119108-100G A119108-10G A119108-25G A119108-1G A119108-5G A119108-500G
103799-166EA 5 USD
103799-166 76995-684 76995-686 76995-688 77297-656 77311-840 77670-680
Boc-Glu(OtBu)-OH 97%
Boc-Glu(OtBu)-OH

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₆
Boiling Pt: 449.8 °C (760 mm Hg)
Melting Pt: 102…105 °C
Flash Pt: 225.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038257
CAS Number: 13726-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR