Boc-Gln(Xan)-OH 97%

Supplier: Ambeed
Warning

A244968-10G A244968-25G A244968-1G A244968-5G
103811-616EA 13 USD
103811-616 103851-450 76976-558 77090-680 77304-342 77304-344
Boc-Gln(Xan)-OH 97%
Boc-Gln(Xan)-OH

Precaución: for Research Use Only

Formula: C₂₃H₂₆N₂O₆
Boiling Pt: 669.8 °C (760 mm Hg)
Flash Pt: 358.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00066158
CAS Number: 55260-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR