Boc-Gln(Trt)-OH 98%

Supplier: Ambeed
Warning

A399486-1G A399486-5G A399486-100G A399486-10G A399486-25G
103827-032EA 6 USD
103827-032 103838-070 76841-510 76841-512 77003-880 77003-882 77307-064
Boc-Gln(Trt)-OH 98%
Boc-Gln(Trt)-OH

Precaución: for Research Use Only

Formula: C₂₉H₃₂N₂O₅
MW: 488.6 g/mol
Boiling Pt: 728.9 °C (760 mm Hg)
Melting Pt: 113…128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153305
CAS Number: 132388-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR