N-Boc-D-valine 99%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Val-OH

A252551-100G A252551-10G A252551-1G A252551-500G A252551-25G A252551-5G
103851-896EA 6 USD
103851-896 76692-540 76692-542 76692-544 76692-546 77091-314 77091-316
N-Boc-D-valine 99%
N-Boc-D-valine

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MW: 217.3 g/mol
Boiling Pt: 341.8 °C (760 mm Hg)
Melting Pt: 164…165 °C
Flash Pt: 160.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00038282
CAS Number: 22838-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR