(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Tle-OH, N-Boc-D-tert-leucine, Boc-D-Tle-O, (R)-2-(tert-Butoxycarbonylamino)-3,3-dimethylbutyric Acid, (R)-2-(Boc-amino)-3,3-dimethylbutyric Acid, Boc-tBu-D-Gly-OH

A209899-10G A209899-1G A209899-25G A209899-5G A209899-100G
103817-964EA 23 USD
103817-964 103848-650 76999-590 77087-010 77087-012 77087-014 77671-862
(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid 95%
(R)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoic acid

Precaución: for Research Use Only

Formula: C₁₁H₂₁NO₄
MW: 231.29 g/mol
Flash Pt: 165.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00065575
CAS Number: 124655-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR